MAN424066520 Manchester Organics

2-Methyl-2-phenoxy-propanoic acid

  • MDL number MFCD00129939
  • Molecular Weight : 180.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H42788
Availability details for
Quantity
  • Availability
  • Add to Cart