MAN424066529 Manchester Organics

2-Methyl-2-(2-methylphenoxy)propanoic acid

  • MDL number MFCD01695483
  • Molecular Weight : 194.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T43027

Safety Information

MSDS https://www.manchesterorganics.com/T43027
Availability details for
Quantity
  • Availability
  • Add to Cart