MAN424066582 Manchester Organics

2-(3-Methoxyphenoxy)-2-methyl-propanoic acid

  • MDL number MFCD03422283
  • Molecular Weight : 210.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A42873

Safety Information

MSDS https://www.manchesterorganics.com/A42873
Availability details for
Quantity
  • Availability
  • Add to Cart