MAN424066634 Manchester Organics

3-Chloropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD02253980
  • Molecular Weight : 197.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G42971

Safety Information

MSDS https://www.manchesterorganics.com/G42971
Availability details for
Quantity
  • Availability
  • Add to Cart