MAN424066645 Manchester Organics

3-(2-Fluorophenyl)-5-methyl-4H-isoxazole-5-carboxylic acid

  • MDL number MFCD07186535
  • Molecular Weight : 223.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U43189

Safety Information

MSDS https://www.manchesterorganics.com/U43189
Availability details for
Quantity
  • Availability
  • Add to Cart