MAN424066650 Manchester Organics

3-(1-Methylpyrazol-4-yl)-4,5-dihydroisoxazole-5-carboxylic acid

  • MDL number MFCD06805565
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z43378

Safety Information

MSDS https://www.manchesterorganics.com/Z43378
Availability details for
Quantity
  • Availability
  • Add to Cart