MAN424066670 Manchester Organics

5-Methyl-2-pyrrol-1-yl-thiophene-3-carboxylic acid

  • MDL number MFCD01108779
  • Molecular Weight : 207.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W43329

Safety Information

MSDS https://www.manchesterorganics.com/W43329
Availability details for
Quantity
  • Availability
  • Add to Cart