MAN424066750 Manchester Organics

2-(5-Amino-4H-1,2,4-triazol-3-yl)acetic acid

  • MDL number MFCD00457974
  • Molecular Weight : 142.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H43087

Safety Information

MSDS https://www.manchesterorganics.com/H43087
Availability details for
Quantity
  • Availability
  • Add to Cart