MAN424066771 Manchester Organics

3-Isobutoxybenzoic acid

  • MDL number MFCD01993662
  • Molecular Weight : 194.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F43108

Safety Information

MSDS https://www.manchesterorganics.com/F43108
Availability details for
Quantity
  • Availability
  • Add to Cart