MAN424066784 Manchester Organics

3-[(4-Chloro-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD01114891
  • Molecular Weight : 236.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X30018

Safety Information

MSDS https://www.manchesterorganics.com/X30018
Availability details for
Quantity
  • Availability
  • Add to Cart