MAN424066808 Manchester Organics

2-Pyrazol-1-ylbenzoic acid

  • MDL number MFCD03086184
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F43131

Safety Information

MSDS https://www.manchesterorganics.com/F43131
Availability details for
Quantity
  • Availability
  • Add to Cart