MAN424066824 Manchester Organics

4-(5-Methyltetrazol-1-yl)benzoic acid

  • MDL number MFCD00458484
  • Molecular Weight : 204.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y43584

Safety Information

MSDS https://www.manchesterorganics.com/Y43584
Availability details for
Quantity
  • Availability
  • Add to Cart