MAN424066838 Manchester Organics

3-(1,3-Dimethylpyrazol-4-yl)isoxazole-5-carboxylic acid

  • MDL number MFCD04970288
  • Molecular Weight : 207.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N43230

Safety Information

MSDS https://www.manchesterorganics.com/N43230
Availability details for
Quantity
  • Availability
  • Add to Cart