MAN424066839 Manchester Organics

2-(4-Bromo-3-cyclopropyl-pyrazol-1-yl)acetic acid

  • MDL number MFCD04970945
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P43231

Safety Information

MSDS https://www.manchesterorganics.com/P43231
Availability details for
Quantity
  • Availability
  • Add to Cart