MAN424066852 Manchester Organics

(1,5-Dimethylpyrazol-4-yl)methanol

  • MDL number MFCD02253731
  • Molecular Weight : 126.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D43175

Safety Information

MSDS https://www.manchesterorganics.com/D43175
Availability details for
Quantity
  • Availability
  • Add to Cart