MAN424066863 Manchester Organics

3-(4-Bromo-3,5-dimethyl-pyrazol-1-yl)propan-1-ol

  • MDL number MFCD06804890
  • Molecular Weight : 233.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R43255

Safety Information

MSDS https://www.manchesterorganics.com/R43255
Availability details for
Quantity
  • Availability
  • Add to Cart