MAN424066875 Manchester Organics

2-([1,2,4]Triazolo[1,5-a]pyrimidin-6-yl)ethanol

  • MDL number MFCD09834256
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D43198

Safety Information

MSDS https://www.manchesterorganics.com/D43198
Availability details for
Quantity
  • Availability
  • Add to Cart