MAN424066889 Manchester Organics

(3-Cyclopropylisoxazol-5-yl)methanol

  • MDL number MFCD16556130
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U43419

Safety Information

MSDS https://www.manchesterorganics.com/U43419
Availability details for
Quantity
  • Availability
  • Add to Cart