MAN424066900 Manchester Organics

(4-Bromo-2-methyl-pyrazol-3-yl)methanol

  • MDL number MFCD18793417
  • Molecular Weight : 191.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F43223

Safety Information

MSDS https://www.manchesterorganics.com/F43223
Availability details for
Quantity
  • Availability
  • Add to Cart