MAN424066907 Manchester Organics

1-[(4-Fluorophenyl)methyl]-3,5-dimethyl-pyrazol-4-amine

  • MDL number MFCD02055632
  • Molecular Weight : 219.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N43299

Safety Information

MSDS https://www.manchesterorganics.com/N43299
Availability details for
Quantity
  • Availability
  • Add to Cart