MAN424066913 Manchester Organics

1-(1,5-Dimethylpyrazol-4-yl)-N-methyl-methanamine

  • MDL number MFCD02055850
  • Molecular Weight : 139.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V43489

Safety Information

MSDS https://www.manchesterorganics.com/V43489
Availability details for
Quantity
  • Availability
  • Add to Cart