MAN424066915 Manchester Organics

1-(1,3-Dimethylpyrazol-4-yl)-N-methyl-methanamine

  • MDL number MFCD02056455
  • Molecular Weight : 139.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X43583

Safety Information

MSDS https://www.manchesterorganics.com/X43583
Availability details for
Quantity
  • Availability
  • Add to Cart