MAN424066926 Manchester Organics

3,5-Dimethyl-1-(p-tolylmethyl)pyrazol-4-amine

  • MDL number MFCD02056373
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J43249

Safety Information

MSDS https://www.manchesterorganics.com/J43249
Availability details for
Quantity
  • Availability
  • Add to Cart