MAN424066931 Manchester Organics

2-Amino-6-ethyl-4,5,6,7-tetrahydrobenzothiophene-3-carbonitrile

  • MDL number MFCD01114964
  • Molecular Weight : 206.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P43323

Safety Information

MSDS https://www.manchesterorganics.com/P43323
Availability details for
Quantity
  • Availability
  • Add to Cart