MAN424066932 Manchester Organics

3-Morpholinosulfonylaniline

  • MDL number MFCD02090909
  • Molecular Weight : 242.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R43324

Safety Information

MSDS https://www.manchesterorganics.com/R43324
Availability details for
Quantity
  • Availability
  • Add to Cart