MAN424066943 Manchester Organics

Methyl 5-amino-4-cyano-3-methyl-thiophene-2-carboxylate

  • MDL number MFCD00126940
  • Molecular Weight : 196.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C43243

Safety Information

MSDS https://www.manchesterorganics.com/C43243
Availability details for
Quantity
  • Availability
  • Add to Cart