MAN424066946 Manchester Organics

4-(3,4-Dichlorophenyl)thiazol-2-amine

  • MDL number MFCD00466323
  • Molecular Weight : 245.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F43269

Safety Information

MSDS https://www.manchesterorganics.com/F43269
Availability details for
Quantity
  • Availability
  • Add to Cart