MAN424066949 Manchester Organics

1-[(4-Fluorophenyl)methyl]pyrazol-3-amine

  • MDL number MFCD02056566
  • Molecular Weight : 191.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J43272

Safety Information

MSDS https://www.manchesterorganics.com/J43272
Availability details for
Quantity
  • Availability
  • Add to Cart