MAN424066960 Manchester Organics

1-(1-Ethyl-3-methyl-pyrazol-4-yl)-N-methyl-methanamine

  • MDL number MFCD02090913
  • Molecular Weight : 153.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W43605

Safety Information

MSDS https://www.manchesterorganics.com/W43605
Availability details for
Quantity
  • Availability
  • Add to Cart