MAN424067097 Manchester Organics

2-Hydroxy-6-methyl-benzoic acid

  • MDL number MFCD01194284
  • Molecular Weight : 152.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D43336
Availability details for
Quantity
  • Availability
  • Add to Cart