MAN424067133 Manchester Organics

3-Methoxy-5-nitro-aniline

  • MDL number MFCD01103799
  • Cas Number : 586-10-7
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/R43370
Availability details for
Quantity
  • Availability
  • Add to Cart