MAN424067168 Manchester Organics

Ethyl 2-amino-5-methyl-thiophene-3-carboxylate

  • MDL number MFCD01922142
  • Cas Number : 4815-32-1
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/D43405
Availability details for
Quantity
  • Availability
  • Add to Cart