MAN424067206 Manchester Organics

4-Amino-1-methyl-pyrazole-3-carboxamide

  • MDL number MFCD03422546
  • Cas Number : 3920-40-9
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-1-methyl-1H-pyrazole-3-carboxamide
Inchi Code 1S/C5H8N4O/c1-9-2-3(6)4(8-9)5(7)10/h2H,6H2,1H3,(H2,7,10)
Inchi Key PKAKWMDYLMOHOG-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/V43443
Availability details for
Quantity
  • Availability
  • Add to Cart