MAN424067254 Manchester Organics

3-Methylpyrrolidin-3-ol

  • MDL number MFCD11100984
  • Molecular Weight : 101.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E43475
Availability details for
Quantity
  • Availability
  • Add to Cart