MAN424067255 Manchester Organics

tert-Butyl (2R)-5-oxopyrrolidine-2-carboxylate

  • MDL number MFCD09261417
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F43545

Safety Information

MSDS https://www.manchesterorganics.com/F43545
Availability details for
Quantity
  • Availability
  • Add to Cart