MAN424067295 Manchester Organics

Methyl 3-amino-1H-pyrazole-5-carboxylate

  • MDL number MFCD09991917
  • Cas Number : 632365-54-9
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-1H-pyrazole-5-carboxylate
Inchi Code 1S/C5H7N3O2/c1-10-5(9)3-2-4(6)8-7-3/h2H,1H3,(H3,6,7,8)
Inchi Key WUKSVVOCYHTIMV-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/F43522
Availability details for
Quantity
  • Availability
  • Add to Cart