MAN424067301 Manchester Organics

4-(1,5-Dimethylpyrazol-4-yl)thiazol-2-amine

  • MDL number MFCD04969956
  • Molecular Weight : 194.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M43643

Safety Information

MSDS https://www.manchesterorganics.com/M43643
Availability details for
Quantity
  • Availability
  • Add to Cart