MAN424067546 Manchester Organics

2-(Difluoromethyl)-1H-benzimidazole

  • MDL number MFCD00463015
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D43842

Safety Information

MSDS https://www.manchesterorganics.com/D43842
Availability details for
Quantity
  • Availability
  • Add to Cart