MAN424067653 Manchester Organics

9H-Fluoren-9-ylmethyl N-[(1S)-1-(hydroxymethyl)-3-methyl-butyl]carbamate

  • MDL number MFCD00235932
  • Molecular Weight : 339.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B43886

Safety Information

MSDS https://www.manchesterorganics.com/B43886
Availability details for
Quantity
  • Availability
  • Add to Cart