MAN424067656 Manchester Organics

1-(1H-Pyrazol-4-yl)ethanone

  • MDL number MFCD09878739
  • Molecular Weight : 110.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E43912

Safety Information

MSDS https://www.manchesterorganics.com/E43912
Availability details for
Quantity
  • Availability
  • Add to Cart