MAN424067675 Manchester Organics

3-Bromo-4-chloro-benzaldehyde

  • MDL number MFCD08059100
  • Molecular Weight : 219.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N44012

Safety Information

MSDS https://www.manchesterorganics.com/N44012
Availability details for
Quantity
  • Availability
  • Add to Cart