MAN424067725 Manchester Organics

2-(P-Tolyl)azepane

  • MDL number MFCD02663755
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/S43923
Availability details for
Quantity
  • Availability
  • Add to Cart