MAN424067765 Manchester Organics

4-(1-Methylpyrazol-4-yl)aniline

  • MDL number MFCD06797404
  • Cas Number : 1079178-22-5
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K43963
Availability details for
Quantity
  • Availability
  • Add to Cart