MAN424067907 Manchester Organics

(4-formylphenyl) benzoate

  • MDL number MFCD00446988
  • Molecular Weight : 226.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N44242

Safety Information

MSDS https://www.manchesterorganics.com/N44242
Availability details for
Quantity
  • Availability
  • Add to Cart