MAN424068004 Manchester Organics

1-Ethyl-5-methyl-pyrazole-4-carbaldehyde

  • MDL number MFCD02056531
  • Molecular Weight : 138.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U44477

Safety Information

MSDS https://www.manchesterorganics.com/U44477
Availability details for
Quantity
  • Availability
  • Add to Cart