MAN424068025 Manchester Organics

5-(Methoxymethyl)furan-2-carbaldehyde

  • MDL number MFCD02253954
  • Cas Number : 1917-64-2
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(methoxymethyl)-2-furaldehyde
Inchi Code 1S/C7H8O3/c1-9-5-7-3-2-6(4-8)10-7/h2-4H,5H2,1H3
Inchi Key ASHVULSQMDWKFO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S44222
Availability details for
Quantity
  • Availability
  • Add to Cart