MAN424068089 Manchester Organics

1-(Benzenesulfonyl)-4-bromobenzene

  • MDL number MFCD00034047
  • Cas Number : 23038-36-0
  • Molecular Weight : 297.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/L44286
Availability details for
Quantity
  • Availability
  • Add to Cart