MAN424068131 Manchester Organics

2-Bromo-3-chloro-phenol

  • MDL number MFCD04037216
  • Cas Number : 855836-62-3
  • Molecular Weight : 207.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J44307
Availability details for
Quantity
  • Availability
  • Add to Cart