MAN424069122 Manchester Organics

Pyrimidine-4,6-diamine

  • MDL number MFCD00044572
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/B45358
Availability details for
Quantity
  • Availability
  • Add to Cart