MAN424069172 Manchester Organics

5-Phenylpyridine-2-carbaldehyde

  • MDL number MFCD10700050
  • Molecular Weight : 183.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F45385
Availability details for
Quantity
  • Availability
  • Add to Cart